Would you like to log in again?
Radiation Lab Activity
Supplier: THE SCIENCE SOURCE SE
16228-01 16225 16226 16226 16228-02
470201-200EA 15.8 USD InStock
470201-200 470023-942 470023-944 470006-730 470002-500
Radiation Lab Activity
Educational Classroom Kits and Activities
Students investigate the absorption and radiation of energy on different surfaces.
  • Activity Includes Light Source
  • Two Experiments in One Activity
  • Students Research Absorption and Radiation of Energy
They then turn on the light at 10 cm distance and observe how temperatures respond differently in the black can and silver can.
In the reverse experiment, the rates of cooling can be determined after students turn off the light.
Ordering information: Each kit includes a 16 oz. black metal and a 16 oz. shiny metal can, two insulating lids, and two other thermometers (-20° to 50°C). The single kit also includes one 120VAC clamp lamp with a reflector and 100 watt light bulb; the class kit includes four lamps.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty