Would you like to log in again?
Due to scheduled system maintenance, vwr.com will be temporarily unavailable from 6:00 am EST to 2:00 PM EST on Saturday, May 25. 
Plant and Animal Mitosis Lab Activity
Supplier: Avantor
470328-422KT 132.19 USD InStock
470328-422 470024-096 470007-014
Plant and Animal Mitosis Lab Activity
Educational Classroom Kits and Activities
Students will gain microtechnique and staining experience by creating their own microscope slides of onion root tips that have been specially grown, cut, and fixed at maximum mitotic activity.
  • Aligned to NGSS standards (MS/HS: LS1)
In the process, they will learn the chronological order of, and the concepts behind, cellular mitosis and division, as well as the differences and similarities between plant and animal cell division. The kit includes materials for 30 students, three reference slides of plant and animal mitosis for comparison, a teacher’s guide, and student copymaster. Note: Coupon included for perishable materials. Redeem by mail, fax, phone, or e-mail.
Due to the nature of this item, it cannot be shipped by air under any circumstances. Please select ground shipping at time of checkout.
The kit is also available for purchase with a link to a NewPath digital lesson, which includes a variety of resources on the topic, including background information, illustrations and self-assessments.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty