Would you like to log in again?
Investigating Transpiration Lab Activity
Supplier: Avantor
WARD470024-118
470024-118KT 215.69 USD InStock
470024-118
Investigating Transpiration Lab Activity
Educational Classroom Kits and Activities
Students use several techniques to investigate the location, methods, and mechanisms of transpiration in plants. Students first determine the source of water loss and confirm their findings with a qualitative chemical test. Then they prepare microscope slides of xylem in stem tissue, and prepare a leaf imprint using an innovative method that vividly highlights stomate structures. Finally, using WARD’S specially-designed potometer and supplied materials, students subject bean seedlings to varying environmental conditions to determine how transpiration is affected. You’ll get enough materials for eight setups, a teacher’s guide, and student copymaster.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty