Would you like to log in again?
Magnetic Fingerprint Kit
Supplier: FORENSICS SOURCE
1-0123
470024-158EA 96.5 USD InStock
470024-158
Magnetic Fingerprint Kit
Educational Materials Forensics Educational Materials Fingerprint Analysis Learning Activities
Includes 90 cc Wide-Mouth Powder Jar for Convenient Use With the Magnetic Applicator.
Professional fingerprinting techniques are easy to complete and duplicate, with this comprehensive kit. Uncover latent fingerprints using a magnetic powder applicator, 1 oz. of bi–chromatic magnetic powder, one 1 1/2" x 360" roll of lifting tape, and one hundred 3" x 5" cards, all contained in a sturdy carrying case. Size: 10 3/8"L x 4 5/8"W x 1 3/4"H.
Available to U.S. customers only; similar product will ship elsewhere.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty