Would you like to log in again?
Ward's® Paper Recycling Kit
Supplier: Avantor
WARD470024-198
470024-198KT 116.99 USD InStock
470024-198
Ward's® Paper Recycling Kit
Educational Classroom Kits and Activities
Increase Awareness of the Waste and Recycling Potential in Your Classroom
  • Students make their own paper from recycled material
  • Includes a teacher's guide
  • Estimated class/lab time required: 30 - 45 minutes plus drying time
Your students will have fun making paper, while at the same time, they will be learning about the costs and benefits of recycling paper. They will keep track of the amount of waste paper generated each day in the classroom and calculate the yearly waste for the classroom, as well as for the entire school. Then, students will make their own recycled paper from waste paper that they collect prior to the lab. By the end of the lab activity, students will be able to consider other materials or methods of recycling. All of the items provided in the kit can be reused. You get enough materials for 15 setups, a teacher’s guide, and student copymaster. Time requirement is 30-45 minutes to create the paper with additional time required for drying paper (will vary according to drying method).
Ordering information: Materials include Pail, 5 qt., Plastic Basin, Plastic Frames, Clips, Spatula, Sponge, Absorbent Pads, Mesh dip net, and Screen sheets (9” x 10”). "Waste” paper, Blender or mixer, Iron (recommended), Bathroom scale, and Container to fit on scale are materials needed but not provided.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty