Would you like to log in again?
Ward's® Fecal Slide Analysis Lab Activity
Supplier: Avantor
WARD470024-214
470024-214KT 194.29 USD InStock
470024-214
Ward's® Fecal Slide Analysis Lab Activity
Educational Classroom Kits and Activities
Identify Parasites and Other Fauna
  • Collect samples from local fauna
  • Includes a teacher's guide
  • Estimated class/lab time required: 45 minutes
Your students will collect fecal matter and then examine it for macrocharacteristics and microfauna. Using basic microbiological techniques, your students will isolate, stain, observe, and identify the bacterial cultures and other parasites. They will then compile a list of bacteria and fungi that inhabit various animals and extrapolate their findings to what they would expect to find in a normal healthy adult human. The activity comes with WARD’S Internal Animal Parasites Poster, enough materials for eight setups, a teacher’s guide, and student copymaster.Time requirement is one 45 minute period.
Ordering information: Lab includes Iodine solution, Sucrose, Microscope slides, Coverslips, WARD’S Internal Animal Parasites poster, WARD’S Agricultural Parasites slide set, Toothpicks, Latex gloves; small, Pipets,Test tubes; 16 x 150 mm, Cheesecloth, Medicine cups, and Paper cones. Fresh feces (less than 24 hours old), Microscope, Water, Resealable plastic bag, Distilled water, Test tube rack, 250 mL beaker, and Scissors are materials needed but not provided.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty