Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Ward's® Gunshot Residue Presumptive Test
Supplier: ALDON CORP SE
DD6002-KIT
470024-240EA 52.59 USD InStock
470024-240
Ward's® Gunshot Residue Presumptive Test
Educational Classroom Kits and Activities
Colorimetric test for trace evidence.
  • Grades 9-12
  • Estimated class/lab time required: 30 minutes
This simple, two-part presumptive test will help you quickly determine whether or not a surface has recently been exposed to a discharged firearm. Simply wipe down the surface in question, add the chemical reagents, and observe the results. A rapid color-change takes place to verify the presence of nitrates, the main component of gunpowder residue (GSR), while a different color change will verify the presence of heavy metals associated with GSR. Each kit contains complete instructions and enough materials to perform 30 tests.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty