Would you like to log in again?
Determination of Amino Acid Sequence in a Peptide Lab Activity
Supplier: AQUAPHOENIX SCIENTIFIC INC. SE
8-160R
470024-300KT 548 USD InStock
470024-300
Determination of Amino Acid Sequence in a Peptide Lab Activity
Educational Materials Biology Educational Materials Biochemistry Learning Activities
Tie Science History to Science Technique
Follow the path that led biochemist Frederick Sanger to receive two Nobel Prizes with this exciting lab. Everything (except a hairdryer) is included for students to determine amino acid content, composition, and position of terminal groups of the tripeptide arginylglycylphenylalanine from a protein hydrolysate. The activity includes an instructor’s manual, an assessment manual, and student data sheets for four setups to do four different exercises each.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty