Would you like to log in again?
Lab-Aids® Investigating Human Respiration Kit
Supplier: LAB-AIDS INC (KITS) SE
803S 803S-RC
470024-332EA 111 USD InStock
470024-332 470024-330
Lab-Aids® Investigating Human Respiration Kit
Educational Materials Biology Educational Materials Human Biology Learning Activities
Explore the role Of respiratory system in regulation of blood gases.
  • Creating models is one way to understand and communicate scientific information
  • Examine human respiration
  • Time required: one to two 50 minutes class periods
Using bromothymol blue (BTB) as an indicator, students conduct a qualitative investigation. The amount of carbon dioxide in their exhaled breath is then quantitatively measured using the indicator to perform a simple titration. The combination of these steps demonstrates the function of the lungs in providing surface area for oxygen to enter and carbon dioxide wastes to leave the blood.
Delivery information: See datasheet for contents information.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty