Would you like to log in again?
Ethidium Bromide
Supplier: ALDON CORP SE
EE0066-0.5ML
470024-556EA 19.7 USD InStock
470024-556
Ethidium Bromide
Chemicals for Science Education
Research-Grade DNA Stain
Visualizes even the smallest bands of DNA. Stain gels in a staining tray for ten minutes with a 1X working solution of ethidium bromide, then decant the ethidium bromide into its original container for reuse (can be reused 7–10 times without substantive loss of staining intensity). Rinse off your gels in a designated sink area, then destain them in water for 10–20 minutes and view gels on a UV transilluminator. The 500 μL at 10mg/mL stock concentrate makes 5 L of 1X working solution. Store indefinitely at room temperature.
Caution: Ethidium bromide is a mutagen. Wear protective gloves and eyewear when handling. Sold only to colleges, universities, and accredited laboratories. Check your local regulations for disposal.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty