Would you like to log in again?
Ward's® Bulk Simulated Blood
Supplier: Ward's
WARD470024-642 WARD470024-640 WARD470024-638 WARD470024-644
470024-638EA 94.5 USD InStock
470024-638 470024-644 470024-642 470024-640
Ward's® Bulk Simulated Blood
Educational Materials Biology Educational Materials Blood Typing Learning Activities
Big enough for all your lab sections.
  • Large Quantities
  • Each Blood Type Provided in 1 L Bottle
  • Convenient Bulk Refills
At the request of our college customers, we now offer these convenient bulk refills for our exclusive Simulated ABO and Rh Blood-Typing Lab Activities.
Instructions for performing blood typing using this product can be found in the teacher's guide and student copy master that accompanied the purchase of the lab activity.
Ordering information: Each blood type is provided in a 1L bottle. This product requires Ward's Simulated Antisera, available separately.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty