Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Buffer Envelopes

Supplier: MICRO ESSENTIAL LABORATORY I
280-PH8 280-PH6 280-PH12 280-211 280-PH11 280-PH3
470009-244PK 42.85 USD InStock
470009-244 470024-668 470024-658 470024-664 470024-676 470024-674
Buffer Envelopes
Buffers Cell Culture Buffers
Shelf Life (months): 36
Storage: Green

Caution: Please Note: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty