Would you like to log in again?
Ward's® Calcite
Supplier: VWR
WARD470024-924
470024-924EA 53.39 USD InStock
470024-924
Ward's® Calcite
Specimens Mineral Specimens Mineral Thin Sections
When minerals, rocks, and fossils cannot be accurately identified by macroscopic observation and testing, thin section slides can reveal the composition and structure of the specimens on a microscopic level.
Thin section slides are precisely ground to a thickness of 0.03 mm across the entire specimen area. Mineral and rock thin sections are mounted on 27 x 46 mm microscope slides, and protected by a glass coverslip. All thin section slides are labeled for reference.
Our excellent series of standard thin sections features important and interesting rock-forming mineral types most often found in field samples.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty