Would you like to log in again?
Ward's® Gneiss (Biotite)
Supplier: VWR
WARD470025-138
470025-138EA 45 USD InStock
470025-138
Ward's® Gneiss (Biotite)
When minerals, rocks, and fossils cannot be accurately identified by macroscopic observation and testing, thin section slides can reveal the composition and structure of the specimens on a microscopic level.
Thin section slides are precisely ground to a thickness of 0.03 mm across the entire specimen area. Mineral and rock thin sections are mounted on 27 x 46 mm microscope slides, and protected by a glass coverslip. All thin section slides are labeled for reference.
Classic North American Rock Thin Sections
Our finest thin sections have been a classroom standard for over 40 years. When purchased individually, each 27 x 46 mm slide is packaged as an Explano-Mount, with a study sheet, in a durable plastic sleeve. In addition, you can now order matching hand specimens for each thin section.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty