Would you like to log in again?
Ward's® Know Your Minerals Collection
Supplier: VWR
WARD470025-164
470328-404KT 77.99 USD InStock
470328-404 470025-164
Ward's® Know Your Minerals Collection
Specimens Mineral Specimens Mineral Collections
Perfect for Independent Study or Class Tests.
  • Aligned to NGSS standards (MS/HS: ESS2, ESS3)
Students learn the techniques of mineral identification by following the instructions for testing the “unknown” specimens in the collection. Specially selected specimens clearly illustrate hardness, cleavage/fracture, tenacity, and streak.
The set is housed in a compartmented collection box, and a comprehensive study guide and identification list is included.
Contents: 22 unnumbered samples, 1×1"; streak plateMinerals:
Hardness: apatite, calcite, fluorite, microcline, quartz, selenite, talc
Clevage/Fracture: calcite, feldspar, halite, mica, quartz (obsidian), serpentine
Tenacity: copper, mica, selenite, sulfur.
Streak: graphite, hematite, magnetite, pyrite, talc.
The kit is also available for purchase with a link to a NewPath digital lesson, which includes a variety of resources on the topic, including background information, illustrations and self-assessments.
Caution: Due to the nature of this product it may contain asbestos.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty