Would you like to log in again?
Introductory Mineral and Rock Collection
Supplier: Avantor
WARD470025-166
470025-166EA 43.9 USD InStock
470025-166
Introductory Mineral and Rock Collection
Specimens Rock Specimens Rock Collections
Examples of the Most Basic Rocks and Rock-Forming Minerals
Give your students a clear introduction to rocks and rock-forming minerals; the specimens in the introductory set are easy to differentiate from one another and clearly show the traits of each type. The set is housed in a compartmented collection box, and an identification key is included.Contents:16 numbered samples, 1" x 1"Minerals: biotite, feldspar, quartz, calciteRocks:Igneous: basalt, granite, rhyolite, gabbroSedimentary: conglomerate, limestone, sandstone, shaleMetamorphic: gneiss, marble, quartzite, schist
Introductory Mineral and Rock Collection, Hand Specimens
The larger hand samples show the features more clearly and are ideal for demonstration or display. The samples come with individual, labelled plastic trays and an identification list.
Contents: 16 numbered samples, 3" x 4"
Introductory Mineral and Rock Thin Section Set
Contents: 16 thin section glass microscope slides, 27 x 46 mm
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty