Would you like to log in again?
Wards® Mohs' Hardness Scale Collection
Supplier: VWR
WARD470025-244
470025-244EA 19.99 USD InStock
470025-244
Wards® Mohs' Hardness Scale Collection
Specimens Mineral Specimens Mineral Collections
Our Most Popular collection.
  • 9 numbered samples
  • Includes streak plate, nail, glass slide, and copper strip
All samples are pure crystalline forms to ensure accurate test results. You can also perform other physical properties tests with the included materials. The set is housed in a compartmented collection box, and an identification list is included.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty