Would you like to log in again?
Ward's® Tenacity Collection
Supplier: VWR
WARD470025-246
470025-246EA 21.09 USD InStock
470025-246
Ward's® Tenacity Collection
Specimens Mineral Specimens Mineral Collections
Five Types of Cohesiveness
Different types of tenacity are shown in five popular minerals. The set is housed in a compartmented collection box, and an identification list is included.Contents:5 numbered samples, 1" x 1"Minerals:Brittle: sulfurSectile: seleniteMalleable: copperFlexible: vermiculiteElastic: mica
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty