Would you like to log in again?
Ward's® Cleavage Collection
Supplier: VWR
WARD470025-248
470025-248EA 22.49 USD InStock
470025-248
Ward's® Cleavage Collection
Specimens Mineral Specimens Mineral Collections
Specimens Illustrate Smooth Mineral Breakage
  • Hand picked by WARD's Geologists
  • Specimen list included
  • Teaches students how minerals natural cleavage
Each sample shows a different type of cleavage that leaves a smooth surface. The set is housed in a compartmented collection box, and an identification list is included.
Ordering information: Contents include 6 numbered samples (1" x 1"), Basal Cleavage (Mica), Pinacoidal Cleavage (Feldspar), Prismatic Cleavage (Amphibole), Cubic Cleavage (Halite), Rhombohedral Cleavage (Calcite), and Octahedral Cleavage (Fluorite).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty