Would you like to log in again?
Fracture Collection
Supplier: VWR
WARD470025-250
470025-250EA 21.59 USD InStock
470025-250
Fracture Collection
Specimens Mineral Specimens Mineral Collections
Demonstrates Irregular Breakage
Each specimen illustrates a type of mineral breakage that does not leave a smooth cleavage surface. The set is housed in a compartmented collection box, and an identification list is included.Contents:6 numbered samples, 1×1"Minerals: Even Fracture: lithographic limestoneUneven Fracture: serpentineConchoidal Fracture: obsidianSubconchoidal Fracture: rose quartzSplintery Fracture: tremoliteHackly Fracture: copper
Caution: Due to the nature of this product it may contain asbestos.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty