Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Introductory Fluorescent Mineral Collection

Supplier: Avantor
WARD470025-300 SK68C-UV
470025-300EA 58.1 USD InStock
470025-300 470349-458
Introductory Fluorescent Mineral Collection
Specimens Mineral Specimens Mineral Collections
Both shortwave and longwave fluorescent minerals included

  • Contains 12 numbered specimens: 6 longwave, 6 shortwave
  • Size: Up to 1×2" per specimen

Fascinate your students as you shine an ultraviolet light to set the fluorescent minerals in this set aglow. The set is housed in a compartmented collection box, and an identification list is included. Contents may vary due to availability.

Ordering information: LED UV Flashlight available to purchase separately. Requires 1 AA battery. UV wavelength 395 nm.

Caution: Due to the nature of this product it may contain asbestos.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty