Would you like to log in again?
Crystal Form Topic Set
Supplier: VWR
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
WARD470025-344
470025-344EA 81.39 USD InStock
470025-344
Crystal Form Topic Set
Specimens Mineral Specimens Mineral Collections
Geometric Shapes and Structures of Crystalline Minerals.
Students can easily learn the primary crystal structures with the samples and activities contained in our complete GEO-logic study set. A teacher's guide and student copymasters are included for the following activities: crystal models, crystal form and habit—measuring crystal faces, and identifying crystal systems. The set is housed in a compartmented collection box.
Contents: 12 numbered samples, 1×1"; magnifier; protractor; ruler Isometric: grossular, pyrite (2) Hexagonal: apatite, corundum, dolomite Tetragonal: vesuvianite, zircon Orthorhombic: aragonite Monoclinic: orthoclase, staurolite Triclinic: microcline.
GEO-logic is a unique, comprehensive series of teaching collections designed to assist both teacher and student in understanding basic earth science concepts through hands-on activities and carefully prepared teacher and student guides. The GEO-logic system provides a modular approach to teaching and allows you to address broad curriculum needs for the study of minerals and rocks; natural resources and energy; and fossils and earth history.
The two types of GEO-logic teaching collections include:
• A Reference Set, which provides an overview of all topics and comes with a teacher's guide
• Activity Sets, which provide greater depth in an individual subject area and include both a teacher’s guide and student copymasters.
The GEO-logic system is:
• Developed jointly by experienced educators and WARD’S staff geologists
• Designed to save valuable planning and preparation time
• Created from quality teaching materials for hands-on learning
• Classroom tested for dependability.
Each GEO-logic collection includes:
• Individually numbered specimens chosen for their quality and character
• Activity guides with learning objectives and hands-on activities for your students to perform
• An attractive, durable compartmented box for easy use and storage.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty