Would you like to log in again?
Ward's® Illite Shale
Supplier: VWR
Avg. Ratings: 5.0 (1 Review)
REVIEWS (1)
Total Ratings: 1
Avg. Ratings: out of 5
Sort all by:
WARD470025-406
470025-406PK 26.55 USD InStock
470025-406
Ward's® Illite Shale
Specimens Rock Specimens Sedimentary Rocks
Green shale which contains approximately 85% Illite. Impurities may include other clay minerals, such as micas and feldspars.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (1)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty