Would you like to log in again?
Ward's® Labradorite (Cleavage)
Supplier: VWR
WARD470025-662 WARD470025-658 WARD470025-660
470025-658PK 42.05 USD InStock
470025-658 470025-660 470025-662
Ward's® Labradorite (Cleavage)
Specimens Mineral Specimens Classroom Minerals
Feldspar group; good cleavages of pure soda lime feldspar; most show twinning and labradorescence.
  • WARD’S offers different specimen sizes to meet your various needs
  • Student specimens, hand specimens, and bulk packs are available
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty