Would you like to log in again?
Ward's® Microfossil Strew Slide Set
Supplier: Avantor
Ward's®
WARD470026-772
470026-772KT 138.49 USD InStock
470026-772
Ward's® Microfossil Strew Slide Set
Educational Materials Earth Science Educational Materials Paleontology Learning Activities
View minute, fossilized life forms
  • Made by Ward's geologists
  • One of a kind paleontology slides
  • Grade for elementary aged to college level students
What may appear to students as grains of sand are really the fossilized remains of marine organisms, including conodonts, forams, molluscs, and more. Samples, from various localities, span several geologic periods. A slide reference list is included. A hand lens or stereomicroscope, available separately, is recommended for best viewing.
Ordering information: Slide set includes 12 numbered cardboard microplanetary slides (1×3").
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty