Would you like to log in again?
Armadillo Skull, Ward’s®
Supplier: SKULLS UNLIMITED
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
SM 010
470027-964EA 163.35 USD InStock
470027-964
Armadillo Skull, Ward’s®
Models Zoology Models
View the special, adapted features of the nine-banded armadillo, Dasypus novemcinctus, that allow it to forage for its insect diet Unmounted for up-close study, it is ideal for comparison with the closely related anteater skull.
  • Natural bone
  • Professionally cleaned and prepared
Ordering information: This is a real specimen, so size and appearance may vary from the image.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty