Would you like to log in again?
Beneficial Insects Riker Mount
Supplier: Riker Mount
R282
470028-064EA 87.5 USD InStock
470028-064
Beneficial Insects Riker Mount
Specimens Preserved Specimens Display Specimens
Teach students that all insects aren’t bad.
Assembled in this collection are beneficial insects that help keep our ecosystem running. Pollinators, perhaps the most beneficial of all insects, are represented by the bumblebee and honeybee. Species predacious on insect pests include the dragonfly, praying mantis, tiger beetle, damselfly, and lady beetle. A carrion beetle, an important scavenger of dead and decaying matter is included. The mount also features insects that are invaluable to aquatic food chains: the water boatman and caddisfly larva. The caddisfly is also an indicator of pollution levels.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty