Would you like to log in again?
Sedimentator™ with Teacher’s Guide
Supplier: HETTICH INSTRUMENTS
00050 & 51 5051
75801-870EA 367.43 USD InStock
75801-870 470029-184
Sedimentator™ with Teacher’s Guide
Models Geology Models
Demonstrate Sediment Deposition in Rivers, Streams, and Lakes
Instead of using static diagrams in textbooks to show the processes of sedimentation and rock formation, let students shake and flip the Sedimentator tube, then watch the materials settle out instantly. Students can predict the order in which different sediments settle, classify the types of sediments, and observe how organic material becomes trapped in sedimentary layers and begins the fossilization process. When the tube is held horizontally and shifted, it stimulates different rates of water flow in a streambed, showing how fast water must flow to move the different types of sediments downstream. Each tube, made of heavy-duty plastic, comes preassembled, so there is setup, no cleanup, no wet sand, and no water mess. It also comes with a student activities booklet and teacher’s guide. Size: 10 1/2" long.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty