Would you like to log in again?
Bone Clones® Hominids Skull Series
Supplier: BONE CLONES, INC.
BH-025 BH-008 WARD470029-238 BH-018 BH-012 BH-023 BH-004 BH-013 BH-021A BH-020 BH-016
470029-220EA 431.1 USD InStock
470029-220 470029-230 470029-222 470029-232 470029-228 470029-216 470029-238 470029-218 470029-224 470029-234 470029-226
Bone Clones® Hominids Skull Series
Models Paleontology Models
The most complete series of human ancestry
  • Buy individually or bundle and save
  • Accurate replicas are great for human biology, evolutionary biology, and athropology courses
Each hominid was carefully researched and re-created in resin from casts that were made from the original fossils, the latest literature, and photographs, then color-coded to distinguish the original fossil remains from the reconstructed parts.
Aegyptopithecus is believed to be an ancestor of Old World monkeys and hominids. Proconsul africanus lived in Africa approximately 14 -23 million years ago. Kenyanthropus was discovered in 1998 near Lake Turkana, Kenya. Dating back 3.5 million years, this hominid challenges Australopithecus afarensis as the direct ancestor of the modern human. A. afarensis is dated at 3.2 million years old. This hominid fossil commonly known as "Lucy" has been considered to be the first human.
A. aetheopicus is considered a prime example of ancient hominid evolution, even though it is not a direct ancestor of humans. A. africanus has been identified as a new, intermediary genus and species of hominid. Homo rudolphensis is contemporaneous and geographically concurrent with H. habilis. H. heidelbergensis (Rhodesian Man) is the first human ancestor found in Africa and is estimated to be about 300,000 years old. H. ergaster ("Nariokotome boy") is the first set of remains dated before 100,000 years that is complete enough to determine brain size and whether or not language was possible. H. erectus ("Java Man") was discovered in 1969 in Indonesia.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty