Would you like to log in again?
Somso® Gorilla Skull
Supplier: MARCUS SOMMER
ZOS 51 ZOS 50
470029-320EA 428.9 USD InStock
470029-320 470029-318
Somso® Gorilla Skull
Models Zoology Models
  • Cast From Real Specimens
  • Male And Female Versions Available For Comparison
  • 5 Year Manufacturers Warranty
Compare the structural differences between male and female gorillas (Gorilla gorilla), noting the smaller overall size, and size of central crest and brow ridges, of the female, by using this unmounted plastic, adult female skull with the SOMSO male gorilla skull. Both feature a removable lower jaw.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty