Would you like to log in again?
3B Scientific® Functional Larynx Model
Supplier: AMERICAN 3B SCIENTIFIC SE
1013870
470029-706EA 345.65 USD InStock
470029-706
3B Scientific® Functional Larynx Model
Models Human Biology Models
Movable Parts Show Action Of Vocal Cords
  • Available In 2 Sizes
  • 3 Year Manufacturers Warranty
  • Includes 3B Smart Anatomy
The arytenoid cartilage, epiglottis, and vocal cords can be manipulated to demonstrate how the larynx moves during speech and respiration, as well as to show the function of the epiglottis during swallowing. The highly detailed, 21/2X-enlarged model is mounted on a base and includes a key identifying seven structures.
Accessories information: For information about 3B Smart Anatomy, please see the document attached on this page.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty