Would you like to log in again?
Denoyer-Geppert® Giant Heart Model
Supplier: DENOYER-GEPPERT SCI CO SE
0101-00
470029-752EA 659.85 USD InStock
470029-752
Denoyer-Geppert® Giant Heart Model
Models Human Biology Models
This two-part model, enlarged 2¹/₂ - 3X, is ideal for group study because of its prominent size and enhanced visibility.
It shows both atria with auricles, the coronary sulcus, the coronary and pulmonary blood vessels, and segments of the esophagus, trachea, aorta, and vena cava.
Students can remove the anterior wall to study the internal structures, including the entrance points of the superior and inferior vena cavae and flexible tricuspid, mitral, and aortic valves.
The model is attached to a base, which represents a portion of the pericardium resting in its natural position on the diaphragm.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty