Would you like to log in again?
Green Hydra Model
Supplier: EDUCATIONAL & SCIEN. PRODUCT
ZKA-300-H
470029-780EA 731.7 USD InStock
470029-780
Green Hydra Model
Models Botany Models
Exceptional Model Of A Commonly Studied Organism
  • Large Model Of Hydra Viridissima
  • Shows General Structure And Budding
  • Includes Keycard
This green hydra model features exceptional detail, including visible nematocysts. Colorfully illustrated and enlarged, the median longitudinal section clearly displays the external and internal structure, budding, spermary and an ovary. This model is mounted on a base.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty