Would you like to log in again?
Somso® Comparative Tapeworm Model
Supplier: MARCUS SOMMER
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
ZOS 116/3
470029-796EA 699 USD InStock
470029-796
Somso® Comparative Tapeworm Model
Models Zoology Models
Comprehensive Review Of Two Types Of Tapeworm
  • Compare 2 Different Species Of Tapeworm In One Model
  • 5 Year Manufacturers Warranty
Compare and contrast the life cycles and structures of Taenia saginata and Taenia solium by studying the display of eight detailed models collected on one base. The models show the scolices, eggs, cysticercus, maturing proglottids, and an actual-size representation of mature proglottids, which is most important to identification.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty