Would you like to log in again?
Rescue Critters® Emily K-9 Positioning Manikin
Supplier: THALES & CO.
300
470029-832EA 5400 USD InStock
470029-832
Rescue Critters® Emily K-9 Positioning Manikin
Models Zoology Models
Featuring Realistic Rotation And Range Of Motion
  • Natural articulation and range of motion
  • Can be adjusted to correct positions for surgery and x-ray
  • Made to order
Students learn how to position dogs for abdominal surgery, x-ray, spay and neuter, advanced spinal stabilization, spinal recumbency and more. It has natural movements and offers resistance; allowing you to safely practice techniques before working on live animals. Fully articulated, Emily has shoulders and hip joints that rotate realistically, with 90° flexibility. Also with 90° range of motion are the knees, elbows, and carpal and hock joints. This baby comes complete with carrying case, kneeling pad and brush.
Ordering information: This item is made to order. Returns cannot be accepted.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty