Would you like to log in again?
Amino Acids SuperModel Kit
Supplier: RYLER ENTERPRISES, INC.
AA-2
470029-876EA 32.65 USD InStock
470029-876
Amino Acids SuperModel Kit
Educational Materials Biology Educational Materials Biochemistry Learning Activities
Teach protein structure with models and activities.
  • Construct all 20 amino acids in both the L and D forms
  • Includes connectors for single and double bonds
  • Instruction booklet
You can construct all 20 amino acids in both the L and D forms, as well as make peptide bonds and polypeptides with these flexible and durable model parts. The kit includes 29 carbons, 16 oxygens, 54 hydrogens, three sulfurs, 11 nitrogens, and 109 connectors for single and double bonds, as well as an instruction booklet.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty