Would you like to log in again?
Basic Buddy® Budget CPR Torsos
Supplier: NASCO HEALTHCARE INC. SE
Basic Buddy®
LF03696 LF03693 LF03697 LF03695 LF03694
470029-890EA 184.45 USD InStock
470029-890 470029-892 470119-824 470119-822 470119-820
Basic Buddy® Budget CPR Torsos
Models Human Biology Models
Buy Just Enough For Your Needs.
  • Includes replaceable lung bags
  • Mouth protection is included on lung bags for sanitary use
  • Available as individual, or bundles of trainers
Inexpensive manikins designed for teaching individuals or large groups the life-saving techniques of CPR. The one-piece, disposable lung/mouth protection bags make this manikin completely sanitary. The airway opens using the head tilt/chin lift method and there is a visible chest rise when ventilated. The xiphoid process provides an anatomical reference point for hand placement and compressions. The manikin also features both adult and child capabilities.
Each trainer comes with 10 lung mouth protection bags. Bundles are also supplied with 1 carrying / storage bag for every 5 Units.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty