Would you like to log in again?
Denoyer-Geppert® Premier Skulls
Supplier: DENOYER-GEPPERT SCI CO SE
SK81 SK80PN
470029-932EA 492.1 USD InStock
470029-932 470121-002
Denoyer-Geppert® Premier Skulls
Models Human Biology Models
Every Premier Skull features individual teeth with complete roots (each in its own socket), not just a set of fused dentures like other synthetic skulls.
Available as a Special edition numbered skull, with a more realistic appearance, or a painted and numbered skull, these skulls have been Repli-cast from a select natural bone specimen for unparalleled realism. An exclusive hinged flap in the right mandible swings open to expose the roots of the two premolars and first two molars in their sockets, plus an impacted third molar (wisdom tooth.) These five teeth are permanently anchored. All the remaining teeth in the upper and lower jaws are secured in their sockets with a permanently-flexible cement which allows you to extract teeth individually for thorough examination, and then reinsert them. Once replaced in their sockets, teeth will not fall out. This unprecedented extraction procedure can be repeated time and again. Detachable springs permit natural articulation of the mandible as well as jaw removal. The sectioned calvaria detaches to access the neuro-cranium.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty