Would you like to log in again?
Denoyer-Geppert® Ultraflex Vertebral Column
Supplier: DENOYER-GEPPERT SCI CO SE
SP69
470029-934EA 395.9 USD InStock
470029-934
Denoyer-Geppert® Ultraflex Vertebral Column
Models Human Biology Models
Show Natural Compression Of Discs
Fitted with discs of ultra soft resilient vinyl, this spine permits maximum manipulative flexibility. The natural movement of the vertebral column is simulated while all bones are featured in accurate detail. Features include the occipital bone of the skull; spinal cord with medulla; vertebral arteries; paired spinal nerves; detachable sacral flap which exposes the branching of the cauda equina; complete pelvic girdle; and movable femur heads. Includes suspension stand. Size: 37"L.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty