Would you like to log in again?
Somso® Flexible Articulated Foot Skeleton
Supplier: MARCUS SOMMER
QS 23
470030-068EA 207.55 USD InStock
470030-068
Somso® Flexible Articulated Foot Skeleton
Models Human Biology Models
Bones Can Be Separated To Reveal Ends
  • Elastic Articulation Allows For Extended Movement
  • All Bones Are Allowed To Move Freely
  • 5 Year Manufacturers Warranty
The articulating surfaces of the foot bones are easy to study with this model. The bones are strung on an elastic cord, which can be stretched to show every aspect of each foot bone. The model includes the distal part of the tibia and fibula.
WARD’S Human Osteological Preparations brochure is available for download.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty