Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Ward's Endospore Staining Kit
Supplier: Ward's
470030-202KT 63.99 USD InStock
470030-202
Ward's Endospore Staining Kit
Educational Materials Biology Educational Materials Microbiology Learning Activities
Examine a non-pathogenic bacterial strain that illustrates endospore formation.
  • Demonstration plate culture of Bacillus cereus
Under environmentally stressful conditions, vegetative bacteria enter a heat-resistant stage that helps them survive; this stage is known as an endospore.
The bacterial culture and stain in this kit provide a striking example of this stage.
The kit includes:
A coupon redeemable for a plate culture of Bacillus cereus.
Blank microscope slides and applicator swabs
Staining solutions
Canada: Containment Level 2 – PHAC License required.
Ordering information: This item contains a coupon for live or perishable materials. Please provide a date that the live or perishable components will arrive.
Delivery information: The kit will ship separately from the live component. Kits do not contain perishable components and generally ship via UPS ground. Perishable components will ship via overnight or second day delivery.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty