Would you like to log in again?
Ward's® Bioremediation of Industrial Pollution Lab Activity
Supplier: Avantor
470030-304KT 138.49 USD InStock
470030-304
Ward's® Bioremediation of Industrial Pollution Lab Activity
Educational Classroom Kits and Activities
Re-create “Pollution” and Observe Bacteria’s Degradation Abilities
  • Linked to a real world scenario
  • Includes a teacher's guide
  • Estimated class/lab time required: 90 minutes over one lab session and three follow-up sessions
Students will be able to study bioremediation, a process that uses naturally occurring organisms to degrade hazardous waste into nontoxic materials, and assess its potential as a viable method of cleaning waste sites with our unique lab activity. In an investigation that spans one 45-minute lab session and three brief, 15-minute follow-up sessions, students grow a naturally occurring bacterial strain and observe its degrading properties on completely safe, simulated “chemical waste”.
Ordering information: Includes enough materials for six setups, a teacher’s guide, and student copymaster. An incubator, required, is available separately. Coupon included for perishable materials. Redeem by phone or e-mail.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty