Would you like to log in again?
Quick-Developing Rotifers
Supplier: FLORIDA AQUA FARMS INC
873120
470030-310EA 28.25 USD InStock
470030-310
Quick-Developing Rotifers
Specimens Live Specimens Aquaria - Living Material
Rotifers are well adapted to adverse conditions; when dry, the eggs remain viable for years.
  • Mixed species
Now you can demonstrate this adaptation by reconstituting the dry eggs supplied. Just add water, and the eggs of different Rotifer species begin to hatch in 24 hours, completing their process in only 72 hours.
These freshwater rotifers grow best on a diet of microalgae and Roti-Rich, available separately. Instructions are included.
Ordering information: This item will ship immediately upon order, unless you specify otherwise.
Delivery information: This item ships directly from our supplier.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty