Would you like to log in again?
Ward's® Termite Trails: Follow The Ink Lab
Supplier: Avantor
470030-316KT 69.29 USD InStock
470030-316
Ward's® Termite Trails: Follow The Ink Lab
Educational Classroom Kits and Activities
An educational and entertaining animal behavior experiment.
  • Allow your students to manipulate animal behavior in the lab
  • No reproductive termites are sent (we do not send queens)
As termites follow the trails drawn by your students, you will discover a fascinating bahavior pattern. This activity utilizes Reticulitermes, a subterranean species of termites, which use trail pheromones to lead fellow members of a colony to a food source.
Ordering information: This item contains a copupon for live or perishable materials.
Ward's has USDA permits on-file to ship termites to most states within the U.S.
Termites cannot ship to Canada.
Delivery information: The kit will ship separately from the live component. Kits do not contain perishable components and generally ship via UPS ground.
Perishable components will ship via overnight or second day delivery.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty