Would you like to log in again?
Ward's® Pond Protist Mixture, Dry Mix
Supplier: VWR
WARD470030-328
470180-620EA 13.95 USD InStock
470180-620 470030-328
Ward's® Pond Protist Mixture, Dry Mix
Specimens Live Specimens Protists - Living Material
Produce a Variety of Organisms Simply by Adding Water
Just add distilled water or aged tap water to our exclusive dry mix and in three to four days you’ll be able to examine a number of protist types. We also include stain and a slowing agent for easier study. The dry mix can be stored for up to two years. It comes with culture materials, DETAIN Protist Slowing Agent, methylene blue stain, instructions, and student copymaster.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty