Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Ward's® Extracting Bacterial DNA Lab Activity
Supplier: Avantor
470030-372KT 169.99 USD InStock
470030-372
Ward's® Extracting Bacterial DNA Lab Activity
Educational Classroom Kits and Activities
Isolate and Study DNA in Minutes
  • Lyse bacterial cells in nothing but water
  • Fast procedure
  • Time Required: 15 minutes
Instead of using diagrams or drawings, students can study the physical characteristics of bacterial DNA right in the lab. With the included macromolecular solution, students compare the similarities between the properties of long, linear-chain macromolecules and those of DNA. Most important, the lab activity can be performed in complete safety since Halobacterium salinarium lyses in water for easy extraction of DNA. The lab activity may be completed in 15 minutes or less. The class size comes with enough materials for six setups. Both sizes include a teacher’s guide and student copymaster.
Ordering information: Coupon is included for redemption of perishable materials. Redeem by mail, phone, fax, or email.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty