Would you like to log in again?
Prokaryotic Cell Model
B160
470031-208EA 595 USD InStock
470031-208
Prokaryotic Cell Model
Models Botany Models
All The Features Common In Bacterial Cells
Enlarged 280,000X, this unique depiction shows the external cellular structures and is cutaway to reveal internal structures. Those structures illustrated include pili, flagella, chromosomal DNA, an inclusion body, plasmids, ribosomes, the cell envelope, and the plasma membrane. It is removable from the stand and comes with a printed key.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty