Would you like to log in again?
Vernier CO₂ Gas Sensor
Supplier: VERNIER SOFTWARE & TECH SE
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Vernier
CO2-BTA
470039-278EA 298.99 USD InStock
470039-278
Vernier CO₂ Gas Sensor
Electrodes Other Electrodes
The CO₂ gas sensor measures gaseous carbon dioxide.
  • Wired connection with BTA plug
The CO₂ gas sensor measures gaseous carbon dioxide in two ranges –0 to 10,000 ppm and 0 to 100,000 ppm. With this sensor, you can easily monitor changes in CO₂ levels occurring in respiration of organisms ranging from peas to humans. A 250 ml respiration chamber with probe attachment is included for running controlled experiments with small plants and animals.
Ordering information: LabQuest® (2, 3, Mini) interface is required but not included.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty