Would you like to log in again?
Student Plasticware Set
Supplier: Shiv Dial Sud & Sons
PLKIT20
470039-358CS 92.15 USD InStock
470039-358
Student Plasticware Set
Educational Classroom Kits and Activities
The assortment of labware contains some of the most commonly used pieces. The plastic set comes with 20 items packed in a plastic storage box.
Delivery information: The set includes 2 Beakers (100 and 250 ml), 2 Graduated cylinders (10 and 100 ml), 1 Funnel, 6 mm top dia., 1 drop dispenser bottle, 3 test tubes, 4 reagent bottles, 1 wash bottle, 6 Pipets.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty