Would you like to log in again?
Porcelain Spot Plates
Supplier: SILICO & CHEMICO PORCELAIN SE
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
J-291 J-289
470039-540EA 99.95 USD InStock
470039-540 470153-506
Porcelain Spot Plates
Staining Tiles
These plates are for use in microchemistry experiments.
  • Glazed Porcelain
  • Acid-Resistant
  • Chemically-Inert
  • Plates Withstand up to 1000°F
The glazed porcelain spot plates are acid resistant and chemically inert.
They can withstand temperatures up to 1000°F.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty